Egg PC ≥95%, Powder

Supplier: AVANTI POLAR LIPIDS INC MS

Synonyms: L-α-phosphatidylcholine (Egg, Chicken), Phosphatidylcholine from egg yolk

131601P5G 131601P10G 131601P1G
100123-352EA 2333.62 USD
100123-352 100120-942 100120-396
Egg PC ≥95%, Powder
Egg PC

L-α-phosphatidylcholine (95%) (Egg, Chicken), powder


Egg PC is a further purification of Total Egg Extract. This Natural Lipid is a mixture, and the structure shown above is only representative of one possible structure present in the product. Refer to chart for average fatty acid distribution.

Storage Temperature: Freezer
CAS Number: 97281-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR