D(+) Maltose, Monohydrate, HiMedia Laboratories
Supplier: HiMedia
RM018-500G RM018-10KG RM018-2.5KG RM018-25KG RM018-5KG
61001-554EA 210.56 USD
61001-554 89229-642 89229-644 89229-646 89229-648
D(+) Maltose, Monohydrate, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Maltose is a sugar. It contains one molecule of water of hydration or is anhydrous used for bacteriological purpose.
  • Apperance: White crystalline powder with a slightly sweet taste
  • Solubility: Freely soluble in boiling distilled water, slightly soluble in alcohol and insoluble in chloroform and ether
  • Clarity: 5% w/v solution in boiling distilled water is clear and colorless