Iodine solution 0.01 N
Supplier: AQUA SOLUTIONS MS
4655-1L
4655-500ML
4655-4L
101223-204EA
45.02
USD
101223-204
101223-148
101227-114
Iodine solution 0.01 N
Iodine
Formula:
I₂ MW: 253.81 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00011355 CAS Number: 7553-56-2 |
Specification Test Results
Actual Normality | 0.0095 - 0.0105 N |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...