Anti-ERK1/2 (phospho-Thr202/Tyr204) Mouse Monoclonal Antibody [clone: 4B11B69]

Supplier: Biolegend
675503 675504
75819-244EA 173 USD
75819-244 75819-246
Anti-ERK1/2 (phospho-Thr202/Tyr204) Mouse Monoclonal Antibody [clone: 4B11B69]
Antibodies
ERK1/2 are members of mitogen-activated kinases (MAPKs) of serine/threonine protein kinases.

ERK1/2 can be activated by a range of extracellular stimuli, such as mitogen, growth factors, neurotransmitters, chemokines, and cytokines, through receptor tyrosine kinases (RTK), G protein-coupled receptors (GPCRs), or protein kinase C (PKC). Upon stimulation, ERK1/2 are phosphorylated by the upstream kinase MEK on residues Thr202 and Tyr204 and in turn phosphorylate many other downstream molecules that are involved in a range of cellular processes such as cell proliferation, differentiation, motility and cell death.

Type: Primary
Antigen: ERK1/2 Phospho
Clonality: Monoclonal
Clone: 4B11B69
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR