Sodium chloride 99,0 ACS, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

S9888-10KG S9888-1KG S9888-2.5KG S9888-20KG S9888-25KG S9888-4X2.5KG S9888-500G S9888-50KG S9888-5KG S9888-6X500G
BJS9888-10KGEA 284.19 USD
BJS9888-10KG BJS9888-1KG BJS9888-2.5KG BJS9888-20KG BJS9888-25KG BJS9888-4X2.5 BJS9888-500G BJS9888-50KG BJS9888-5KG BJS9888-6X500G
Sodium chloride 99,0 ACS, Fluka™
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Density: 2.160 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Assay Min. 99.0 %
pH (5 %, 25 °C) 5.0 - 9.0
Water insoluble matter Max. 0.005 %
Barium (Ba) complying
Calcium (Ca) Max. 0.002 %
Iron (Fe) Max. 2 ppm
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.001 %
Heavy metals (ICP) ACS Max. 5 ppm
Bromide (Br) Max. 0.01 %
Chlorate, Nitrate (as NO₃) Max. 0.003 %
Iodide (I) Max. 0.002 %
Phosphate (PO₄) Max. 5 ppm
Sulfate (SO₄) Max. 0.004 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR