Potassium iodide solution 20% (w/v)

Supplier: AQUA SOLUTIONS MS
Warning

7401-1L 7401-4L 7401-20L 7401-500ML
101204-060EA 127.34 USD
101204-060 101203-994 101203-930 101204-610
Potassium iodide solution 20% (w/v)
Potassium iodide
Formula: Kl
MDL Number: MFCD00011405
CAS Number: 7681-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR