Acetonitrile 25% (v/v) in water
Supplier: AQUA SOLUTIONS MS
|
Danger
|
Synonyms:
Methane, cyano-
, Methyl cyanide
, Ethanoic acid nitrile
, Methanecarbonitrile
, Ethanenitrile
, Cyanomethane
, ACN
M-521
102853-450EA
284.07
USD
102853-450
Acetonitrile 25% (v/v) in water
Acetonitrile
Formula:
H₃CCN MW: 41.05 g/mol |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 UN: 1648 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...