Ammonium molybdate(VI) 4% (w/v) in aqueous solution
Supplier: AQUA SOLUTIONS MS
Synonyms:
Molybdic acid ammonium salt, Molybdic acid, ammonium salt, Ammonium molybdate, di-Ammonium molybdate
0458-1L
0458-500ML
101204-150EA
91.51
USD
101204-150
101203-590
Ammonium molybdate(VI) 4% (w/v) in aqueous solution
Ammonium molybdate(VI)
Formula:
H₈MoN₂O₄ |
MDL Number:
MFCD00011596 CAS Number: 13106-76-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...