Principles of Gel Filtration Chromatography, Edvotek
Supplier: EDVOTEK
108
470311-164EA 148 USD
470311-164
Principles of Gel Filtration Chromatography, Edvotek
Educational Classroom Kits and Activities
Introduce chromatography by separating dyes on the basis of size and shape.
  • For ten lab groups
  • Instructions included
  • Packing column: 20 minutes; column separation: 40 minutes
Introduce chromatographic separation to your class and show them how dyes of different colors separate based on their size and shape. This experiment contains materials for dye separation which include dye sample, elution buffer and plastic disposables. Columns may be rinsed and reused.
Storage: room temperature.
Ordering information: Requires 50 or 100 ml beaker or test tube, ring stand with clamps, distilled water.
Delivery information: Kit includes: instructions, sample mixture, chromatography columns, dry matrix, elution buffer, transfer pipets, micro test tubes.