Water USP for injection (WFI), sterile
Supplier: BAXTER IV SOLUTIONS
Synonyms:
Dihydrogen oxide
2B0304X
2B0304X
80089-742CS
1016.1
USD
80089-742
BAL80089742BFM
Water USP for injection (WFI), sterile
Water
Sterile water for injection. For drug diluent use.
- Specific gravity: 1
- Fill range volume (ml): 1030 to 1070
- Store at room temperature (25 °C)
- pH 5.5
- NDC number: 0338-0013-04
Formula:
H₂O MW: 18.02 g/mol |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...