1,2-Distearoyl-sn-glycero-3-phosphocholine ≥99% (by TLC)
Supplier: 0000018221
Synonyms:
1,2-Distearoyl-sn-glycero-3-phosphorylcholine
, DSPC
, 1,2-Dioctadecanoyl-sn-glycero-3-phosphocholine
, (R)-2,3-Bis(stearoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate
, (R)-2,3-Bis(stearoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate
850365P1G
850365C1G
850365C25MG
850365P25MG
850365C500MG
850365P200MG
850365C200MG
850365P500MG
100126-512EA
147.69
USD
100126-512
100122-202
100121-652
100126-646
100125-178
100123-996
100128-924
100122-998
1,2-Distearoyl-sn-glycero-3-phosphocholine ≥99% (by TLC)
1,2-Distearoyl-sn-glycero-3-phosphocholine
Purified by HPLC.
Formula:
C₄₄H₈₈NO₈P MW: 790.16 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00036905 CAS Number: 816-94-4 |
Specification Test Results
TLC, Purity |
|
TLC, Ninhydrin | Negative |
TLC, Iodine | One Major Spot |
TLC, Phosphorus | Positive |
TLC, Charring | Negative |
TLC, Water Dip | One Major Spot |
FAME (GC/FID), NLT 99% | 18:0 (AUC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...