Brain Extract Polar, Avanti Polar Lipids
Supplier: AVANTI POLAR LIPIDS INC MS
141101P500MG 141101C500MG 141101C100MG 141101P100MG
100126-840EA 1015.38 USD
100126-840 100127-072 100123-320 100122-630
Brain Extract Polar, Avanti Polar Lipids
Protein Extraction Protein Extraction Kits
Polar lipid extract is derived from the total lipid extract by precipitation with acetone followed by extraction of the acetone insoluble material with diethyl ether.
  • Hygroscopic
  • Stability: 1 year
  • Storage temperature: –20 °C