Potassium chromate 63% (w/v)

Supplier: AQUA SOLUTIONS MS

7040-500ML
101203-842EA 238.29 USD
101203-842
Potassium chromate 63% (w/v)
Potassium chromate
Formula: K₂CrO₄
MW: 194.19 g/mol
MDL Number: MFCD00011368
CAS Number: 7789-00-6
UN: 3139
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR