(3-Mercaptopropyl)trimethoxysilane ≥98.5%

Supplier: CHEM IMPEX INTL INC MS

Synonyms: Trimethoxy(3-mercaptopropyl)silane, 3-(Trimethoxysilyl)propane-1-thiol, 3-(Trimethoxysilyl)-1-propanethiol

26923-1KG 26923-25G 26923-100G 26923-250G 26923-25G
102570-316EA 392.28 USD
102570-316 102584-472 102627-160 102627-162 CH26923-25G
(3-Mercaptopropyl)trimethoxysilane ≥98.5%
(3-Mercaptopropyl)trimethoxysilane
Formula: HSH₂CCH₂CH₂Si(OCH₃)₃
MW: 196.34 g/mol
Boiling Pt: 215
Melting Pt: -50
Flash Pt: 95.55 °C (204 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00004901
CAS Number: 4420-74-0
UN: 3082
ADR: 9 III

Order Now

Specification Test Results

Assay ≥98.5% (TIPT)
Acidity 6.0 - 7.0 pH
Specific Gravity 1.057 ± 0.0050
Refractive Index 1.440 ± 0.005
Free Acid 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR