Formalin 10%, buffered
Supplier: AVANTIK MS
RS4064
RS4061
MSPP-RS4064EA
665.12
USD
MSPP-RS4064
MSPP-RS4061
Formalin 10%, buffered
Formalin
This 10% Neutral Phosphate Buffered Formalin is a ready-to-use fixative.
The cubitainer packaging comes with a convenient carry handle and dispensing spout.
Formula:
CH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...