Kanamycin sulfate, Ultra Pure Grade

Supplier: VWR
Danger

Synonyms: Kanamycin Monosulphate(2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphateKanamycin AKanamycin Monosulfate(2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulfate

VWRV0408-EU-10G VWRV0408-EU-25G
75856-686EA 161.25 USD
75856-686 75856-684
Kanamycin sulfate, Ultra Pure Grade
Kanamycin sulphate

An aminoglycoside antibiotic that inhibits protein synthesis in gram-negative and gram-positive bacteria and in mycoplasma. Recommended working concentration: 30µg/mL.

Formula: C₁₈H₃₈N₄O₁₅S
MW: 582.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070253
CAS Number: 25389-94-0

Order Now

Specification Test Results

Potency (on anhydrous substance) Min. 750 μg/mg
Potency 0 - 9999 μg/mg
Chromatographic purity Passes test
Crystallinity Passes test
Identification Passes test
pH (25°C; 1 %) 6.5 - 8.5
Loss on drying Max. 4.0 %
Residue on ignition Max. 1.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR