Hand Soldering Tweezer Cartridge, Blade, OK International
Supplier: OK International
TFP-BLP1 TCP-BLP1 TCP-BLH50
76245-658EA 228.81 USD
76245-658 76253-542 76245-642
Hand Soldering Tweezer Cartridge, Blade, OK International
Soldering Supplies Soldering Tips
Precision tweezer cartridge used with the MFR single and dual platforms (MFR-1100 and MFR-2200) with the MFR-H4-TW tweezer hand-piece. Removes even the smallest of discrete components, such as 0201 packages. Second digit denotes substrate material (damage tolerance).