Micro-Lite® High Intensity LED Ring Illuminator, O.C. White
Supplier: O.C. White
Micro-Lite®
LV3500 LV2000-B LV3500REP
76242-506EA 1101.17 USD
76242-506 76241-014 76247-642
Micro-Lite® High Intensity LED Ring Illuminator, O.C. White
Lights LED Lights, Lamps and Bulbs
This LED illuminator is the brightest, and rivals 150 W halogen illuminators with the same yellowish light color.
  • Total energy consumption of this unit is a scant 8 W
  • Features auto switchable transformer design
  • Black molded shades, ESD safe