6-Aminocaproic acid 98.5-101.5%, white crystalline powder USP
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
ε-Amino-n-caproic acid
, H-Eahx-OH
, Aminocaproic Acid
, 6-Aminocaproic Acid
, 6-Aminohexanoic acid
02556-100G
02556-250G
02556-1KG
02556-5KG
102617-450EA
74.98
USD
102617-450
102565-468
102589-400
102562-318
6-Aminocaproic acid 98.5-101.5%, white crystalline powder USP
6-Aminocaproic acid
Formula:
C₆H₁₃NO₂ MW: 131.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00008238 CAS Number: 60-32-2 |
Specification Test Results
Appearance | White crystalline powder |
Heavy Metals | ≤20 ppm |
Infrared Spectrum | Conforms to Structure |
Loss on Drying | ≤0.5% |
Residue On Ignition | ≤0.1% |
Assay | 98.5 - 101.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...