Acetonitrile-[D₃] ≥99.5%

Supplier: CAMBRIDGE ISOTOPE LABS INC MS
Danger

Synonyms: Acetonitrile-d3Cyanomethane-D3Ethannitrile-D3Methyl-d3 cyanideAcetonitrile-[²H₃]Methyl cyanide-D3Trideuteroacetonitrile

DLM-21-1000 DLM-21-10 DLM-21-10X0.75 DLM-21-10X1 DLM-21-50 DLM-21-25
101322-076EA 5817.92 USD
101322-076 101322-098 101322-074 101322-100 101322-078 101322-080
Acetonitrile-[D₃] ≥99.5%
Acetonitrile-[D3]
Formula: D₃CCN
MW: 44.03 g/mol
MDL Number: MFCD00001881
CAS Number: 2206-26-0
UN: 1648
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR