AURORA™ Surgical Gowns, Medline Industries
Supplier: Medline
DYNJP2702
76267-826CS 674.81 USD
76267-826
AURORA™ Surgical Gowns, Medline Industries
Disposable Garments
The choice for a high level of AAMI Level 3 and AAMI Level 4 protection. The comfort and protection are important traits in a surgical gown, especially during extended procedures. But most gowns require a compromise of one quality or the other. Aurora surgical gowns are the exception. They combine both exceptional barrier performance and superb breathability, giving surgeons the best of both worlds.
  • Breathable and comfortable
  • Optional raglan cut
  • Advanced material quickly transfers heat away, keeping the wearer cool and comfortable
  • Roominess and a larger sweep makes movement easier