Ultrabake™ UWM-SH Silicone Washer Pull Plugs and Masks, Caplugs
Supplier: Caplugs
UWM-128 UWPP-M4LS
76248-264PK 174.08 USD
76248-264 76247-476
Ultrabake™ UWM-SH Silicone Washer Pull Plugs and Masks, Caplugs
Fittings and Connectors
Ultrabake™ Silicone Pull Plugs Long series are designed to mask plain and threaded holes; they will mask off the hole and leave a circular grounding or earthing area around the hole. Recommended for powder coating, e-coating, plating and anodizing.
  • For High-temperature processes up to 600 °F (315 °C)
  • Washer design eliminates the need for two-stage masking
  • Washer is dual-purpose designed to be used with counter sunk holes to mask off the faces of the countersink
  • Long series for harder to reach areas
UWM-SH Ultrabake™ Washer Masks are designed to cover plain and threaded studs. These washers are to be used with the UWPP-SH and UWPPLS-SH Series. They will mask off the stud and allow finishing up to the stud.