NASE-1C Two Tools Nano Rework Station, JBC
Supplier: JBC
NASE-1C
76268-272EA 5531.74 USD
76268-272
NASE-1C Two Tools Nano Rework Station, JBC
Soldering Supplies Soldering Stations
The best solution for reworking very small-sized components requiring the highest precision such as chips 0402, 0201, 01005.
  • Comes with the new temperature dynamic profiles functionality
  • Operating temperature: 10 to 40 ºC
  • Robot connector: RJ12 for RS-232 protocol
  • Communication connectors: USB-A/USB-B/Pedal
It works simultaneously with the NT115-A nano handpiece and AN115 steady nano tweezers. The use of these profiles allows you to control the temperature of the component during all the phases of the soldering process, emulating the heating process in a reflow oven.