Solder Workstand with Auto Sleep, OK International
Supplier: OK International
WS1
76245-392EA 282.99 USD
76245-392
Solder Workstand with Auto Sleep, OK International
Soldering Supplies Desoldering Tools
Solder workstand for MFR series.
  • Auto sleep function helps save tip/cartridge life
  • Comes with yellow sponge and brass pad to clean the tip/cartridge
Compatible with MFR-H1-SC2, MFR-H2-ST2, MX-RM3E and MX-H1-AV hand-pieces.