Hematoxylin, OPTIK Type 2

Supplier: Avantik

Synonyms: (6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaolHydroxybrasilinNSC 2785DcH-48Hydroxybrazilin(+)-HematoxylinNatural Black I

RS4364-A
76270-176CS 990.45 USD
76270-176
Hematoxylin, OPTIK Type 2
Hematoxylin

Optik Type 2 Hematoxylin is a progressive stain producing crisp, well delineated chromatin while eliminating background staining.


  • Has no affinity for mucin staining.
  • Contains aluminum salt mordents which may form small, harmless crystals on the bottom of the bottle.
  • These crystals will not interfere with staining and, if troublesome, can be filtered out.
  • Type 2 hematoxylin contains no mercury or alcohol.

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR