Hematoxylin, OPTIK type 1
Supplier: Avantik
Synonyms:
(6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaol
, Hydroxybrasilin
, NSC 2785
, DcH-48
, Hydroxybrazilin
, (+)-Hematoxylin
, Natural Black I
RS4358-A
76270-164CS
893.42
USD
76270-164
Hematoxylin, OPTIK type 1
Hematoxylin
A uniquely formulated progressive Hematoxylin that will produce crisp, well delineated chromatin while eliminating background staining.
- Exceeds Gills and Harris.
- Will not form surface sheen or require daily filtering.
- Contains no mercury or alcohol.
- Hematoxylin DK provides a darker, more intense stain.
Formula:
C₁₆H₁₄O₆ MW: 302.28 g/mol |
MDL Number:
MFCD00078111 CAS Number: 517-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...