Aqueous Clarifier - Optik Type 2, Avantik
Supplier: Avantik
RS4366-B
76270-180CS 1000.46 USD
76270-180
Aqueous Clarifier - Optik Type 2, Avantik
Stains Microscopy and staining reagents
Optik Type 2 Aqueous Clarifier is designed to be used with Type 2 Hematoxylin to produce cell transparency and remove any non-specific staining. Slightly stronger clarifier than the Optik Alcoholic Clarifier.