Optik Bluing Reagent
Supplier: Avantik
RS4362-A RS4363-B RS4368-B
76270-172CS 543.07 USD
76270-172 76270-174 76270-184
Optik Bluing Reagent
Stains Microscopy and staining reagents
Optik Type 1 Bluing Reagent is bluing reagent with a pH of 8.0 to assure proper bluing of hematoxylin stained tissue sections. It is also more resistant to the effects of carry-over and remains in the proper pH range for a longer period of time.
  • Assures proper bluing of hematoxylin stained tissue sections.
  • More resistant to the effects of carry-over.
  • Remains in the proper pH range for a longer period of time.
  • Available in a concentrated solution to be mixed or in a convenient, ready-to-use gallon.
Optik Type 2 Bluing reagent is buffered with a pH of 8 to assure bluing of hematoxylin stained tissue sections.
Supplied in a ready to-use formulation.