Alcohol reagent ≥95% ACS
Supplier: Avantik
Synonyms:
Reagent alcohol
RS4031
76270-154CS
124
USD
76270-154
Alcohol reagent ≥95% ACS
Alcohol
A.C.S. grade. Meets and
exceeds histological grade.
Avantik Alcohol is made from A.C.S. Grade Reagent Alcohol. The lower alcohol content varieties are made with A.C.S. Grade 100% Alcohol. The highest quality for all histology processing and staining procedures.
MDL Number:
MFCD00003568 CAS Number: 64-17-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)