700 System Premium Piston, OK International
Supplier: OK INTERNATIONAL TE
7100009WPK 7300009WPK 7050006RPK
76239-344PK 68.02 USD
76239-344 76239-394 76239-874
700 System Premium Piston, OK International
Fluid Adhesive Dispensers and Pistons
Tight fit Wiper pistons (white) provide a complete seal for low viscosity fluid. It is made from polyethylene