PureFlo® PTFE Mini Filter Cartridge, Zenpure Industries
Supplier: ZENPURE AMERICAS C/O ST GOBAIN TE
MMF020FLO2P
76241-276PK 2489.62 USD
76241-276
PureFlo® PTFE Mini Filter Cartridge, Zenpure Industries
Filters Filter Cartridges
PureFlo® PTFE mini filter cartridges are highly retentive, hydrophobic PTFE membrane filters that have been specially designed for critical applications. No adhesives, binders, or surfactants are used in the manufacturing process. All cartridges are rinsed with high-purity pyrogen-free water to reduce extractables and downtime.
  • Buffers and media
  • Fermentation tanks
  • Scale up processing
  • Gas filtration
The naturally hydrophobic PTFE membrane is ideally suited for filtering aggressive chemicals such as acids, bases, and solvents. In addition, the hydrophobic
membrane provides superior flow and pressure drop characteristics per unit area for gas filtration and tank venting applications. All filter cartridges are 100%
integrity tested to ensure filter performance each and every time out of the package. We are currently offering seven different PTFE pore sizes with eight
different end modifications and three different length options to meet the needs of a wide variety of processes.