700 System Premium Syringes, Ok International
Supplier: OK International
7550LL1NPK 7100LL1NPK 7550LL1DPK 7030LL1NWPK 7050LL1NBLPK 7300LL1NPK TE-15LL-N 7550LL1NWPK
76239-350PK 119.87 USD
76239-350 76239-340 76240-562 76242-224 76239-850 76239-400 76239-334 76239-610
700 System Premium Syringes, Ok International
Syringes General-Purpose Syringes
Ultra-low draft construction of inner diameter yields high accuracy and stability.
  • Silicone and chloride free
  • Low friction polypropylene
Available in three different colors: Natural for most generic applications, amber provides protection of UV/visible light block (up to 520nm), but with the ability to see material inside the barrel; and black for total light block.