Chicago Latrobe® NAS-Type Heavy-Duty Jobber Length Drills, Cutting Tool Supply
Supplier: Cutting Tool Supply
41737 41683
76253-666PK 182.32 USD
76253-666 76250-282
Chicago Latrobe® NAS-Type Heavy-Duty Jobber Length Drills, Cutting Tool Supply
Tool Sets
Heavy-duty construction for drilling in tougher materials and 135° P3 split point is self-centering for reduced thrust and easier penetration.
  • Made of premium high-speed steel
Black oxide surface finish increases wear resistance and adds lubricity to improve chip flow.
Certifications: Manufactured to NAS 907 Type B geometry aerospace specifications.