Chicago Latrobe® Solid Carbide Stub Length Drills, Cutting Tool Supply
Supplier: Cutting Tool Supply
78680 78679 78669
76250-412EA 93.93 USD
76250-412 76250-418 76250-416
Chicago Latrobe® Solid Carbide Stub Length Drills, Cutting Tool Supply
Tool Sets
These drills are manufactured from solid submicron grain carbide.
  • 118° 4-facet point
  • Stub length for rigid setups
  • Uncoated drills standard from stock
These drills are featuring with two flutes, right-hand spiral and right-hand cut.