New methylene Blue N Brecher formulation, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Basic Blue 24Methylene Blue N

Management of Change (MOC) category = NI

BAKRR769-01 BAKRR769-01
JTR769-1CS 2007.67 USD
JTR769-1 CAJTR769-1
New methylene Blue N Brecher formulation, J.T.Baker®
New methylene Blue N

Formulation: 5.0 g/L New Methylene Blue N, 16.0 g/L potassium oxalate, monohydrate in water.


Caution: For R and D use only. Not on TSCA Inventory.

Formula: C₁₈H₂₂ClN₃S
MW: 347.91 g/mol
Density: 1.10 g/cm³
MDL Number: MFCD00135878
CAS Number: 1934-16-3

Order Now

Specification Test Results

For R&D use only. Not on TSCA Inventory.
Absorbance of a 1500 Water Solution (10-mm path vs Water) - 632 nm 0.450 - 0.720
Absorbance of a 1500 Water Solution (10-mm path vs Water) - 285 nm 0.300 - 0.450
Reticulocyte Staining Characteristics Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR