Static Dissipative and Conductive Protectors for D-Subminiature Connectors, DCC Series, Caplugs
Supplier: Caplugs
DCC-02C DCC-05SD
76241-948PK 4022.62 USD
76241-948 76244-772
Static Dissipative and Conductive Protectors for D-Subminiature Connectors, DCC Series, Caplugs
Electrical Sockets
DCC Series Caplugs are designed to provide electrostatic discharge protection of rectangular D-Subminiature input/output type connectors. Molded with an integral flange to enable easy application and removal, Series DCC is available in either black conductive polyethylene or pink static dissipative ethylene vinyl acetate.