Test Lead Set, Newark/Element
Supplier: Newark
5309A
76251-834EA 79.63 USD
76251-834
Test Lead Set, Newark/Element
Cables
DMM Test Lead Kit with probe body and banana plug. Probe body incorporates a two step mold process. The probe with Santoprene® rubber outer insulation, polypropylene fire retardant inner insulation, stainless steel tip, 18AWG tinned copper strand silicone insulation 0.144" outer diameter wire.
  • High temperature resistance
  • 1000Vrms Voltage rating
The banana plug with nickel-plated brass body, nickel-plated beryllium copper spring, polypropylene insulation. Designed for DMM's with non-sheathed banana jacks. Probe body and banana plug are molded directly to the wire for robust pull strength.