Prolite® Polypropylene Tubing, Newage Industries
Supplier: NewAge Industries
448-0147-100
76238-198EA 143.45 USD
76238-198
Prolite® Polypropylene Tubing, Newage Industries
Tubing Transfer Tubing
Prolite is made from a compound that exhibits good heat aging resistance. It meets the FDA requirements for the safe use of olefin polymers used to make articles intended for food contact.
  • Polypropylene homopolymer made from ingredients that meet FDA requirements for food contact surfaces
  • Animal-free ingredients
  • No phthalates used in the manufacture of its ingredients
  • Extremely lightweight, thermoformable and weldable
  • Very low moisture absorption
  • Excellent chemical resistance
  • Naturally translucent white in appearance
  • May be used as a lower-cost alternative to fluoropolymer tubing
It is frequently used in food processing and other industries for clean applications. It is a naturally white, semi-rigid product offering limited ability to bend. Since it is weldable.
Polypropylene is the lightest of all thermoplastic materials. It offers high strength- to-weight ratio, low surface friction, and excellent resistance to corrosion.