Dri-Shield® Moisture Barrier Bag, 2000 Series, SCS Division of Desco, Desco Industries
Supplier: Desco
7002436 7001010
76241-598PK 630.46 USD
76241-598 76245-154
Dri-Shield® Moisture Barrier Bag, 2000 Series, SCS Division of Desco, Desco Industries
Bags Sample Bags
This bag is designed for moisture and electrostatic discharge (ESD) sensitive items.
  • Protects contents from ESD charges and discharges
Allows bag to be grounded when placed on a grounded surface or handled by a grounded operator.
Certifications: Meets ANSI/ESD S20.20, ANSI/ESD S541, EIA 541, JESD625-A, and EIA 583.