Desoldering Cartridges, STDC, OK International
Supplier: OK International
STDC-104 STDC-105 STDC-703L STDC-103 STDC-804L STDC-107
76242-812EA 161.22 USD
76242-812 76241-512 76241-510 76243-380 76241-508 76242-560
Desoldering Cartridges, STDC, OK International
Soldering Supplies Desoldering Tools
Compatible with MX-500 and MX-5000 Systems, MX-DS1 Desoldering hand-piece.