Argyle™ Intermittent Infusion Plug, Cardinal Health
Supplier: Cardinal
8888115006
60351-100BX 85.76 USD
60351-100
Argyle™ Intermittent Infusion Plug, Cardinal Health
Infusions
The infusion plug has a male luer lock design which also accomodates luer slip fitting.
  • Sterile
  • Special synthetic diaphragm retained by secure plastic shrink sleeve
  • Protective sleeve over connector end
Packaging: 50 box/250 case.