SMD Parallel Remover Handpiece, American Hakko Products
Supplier: American Hakko Products
FM2022-02
76248-238EA 726.41 USD
76248-238
SMD Parallel Remover Handpiece, American Hakko Products
Soldering Supplies Soldering Irons
Parallel Remover uses revolutionary parallel action between the tip ends-one tip remains stationary allowing greater surface contact between the tip and the component and providing more efficient and even heat transfer.
  • Parallel Remover uses the same quick-change composite tip technology
  • Fixing collar properly aligns the tips opposite each other
  • Changing tips for different types of components
Removing small chips or larger SOP components, the parallel action of the FM-2022 also allows for a more secure hold onto the component.