Polyester Film Tape 850, 3M
Supplier: 3M
7100077066 7000035828 7100065924 7100065924 7000123285
76242-718CS 8203.21 USD
76242-718 76242-720 76243-326 76243-644 76268-322
Polyester Film Tape 850, 3M
Tapes
Polyester Film Tape 850 is generally used for splicing and web applications. The backing is a thin, high-strength polyester which is abrasion resistant to help reduce splice failures while resisting breakdowns during rough handling and web processing.
  • Low modulus, low Shore A Hardness
  • Thin, high strength polyester film
  • Aggressive acrylic adhesive
Coated with an aggressive pressure sensitive acrylic adhesive, this tape grabs on contact and holds securely. It meets low-caliper requirements to ensure it won't interfere with most machinery. The tape can withstand a wide temperature range from –60 °F/–50 °C to 300 °F/150 °C.