Excellence XPR Legal for Trade (NTEP) Precision Balances, METTTLER TOLEDO®
Supplier: Mettler Toledo
30316508 30316543 30316528 30316480 30315627 30315651 30316566 30317126 30317404 30317442 30317452 30317461 30317436 30317500 30317494 30317486
76098-028EA 12132.5 USD
76098-028 76098-030 76098-032 76098-090 76098-100 76098-102 76098-114 76098-120 76098-122 76098-124 76098-126 76098-128 76098-132 76098-134 76098-136 76098-140
Excellence XPR Legal for Trade (NTEP) Precision Balances, METTTLER TOLEDO®
Balances and Scales
XPR balances deliver accurate results, outstanding weighing performance and support the highest requirements for data integrity and compliance. Quality assurance features ensure your weighing results are always valid, helping you to comply with even the strictest regulations.
  • Made of chemically resistant materials
  • 7" color capacitive touchscreen terminal, glove compatible
  • Storage of weighing methods (including sample series and tolerances)
  • Levelcontrol: level warning with graphical leveling guide
  • Easy connectivity and data export: 4×USB, 1 LAN
  • Integrated label editor, alphanumeric and barcode
  • Export of process and results data to XML or Excel
  • Integrated overload protection
  • Designed for easy cleaning
LabX automatically saves all results and process information securely in a central database. This ensures traceability and assists with FDA 21 CFR part 11 compliance.
The extraordinary stability provided by the new Monobloc™ weighing cell as well as the rugged balance design means you get your results fast even under challenging production environments.
Offer easy connection of peripheral devices like printers, PC, barcode readers, footswitches and sensors. Designed with temperature and time programmable automatic adjustment (proFACT). With customizable tolerance profiles and minimum weigh warning function.