DR560 Desoldering Iron, JBC
Supplier: JBC
DR560-A
76245-648EA 1106.02 USD
76245-648
DR560 Desoldering Iron, JBC
Soldering Supplies Desoldering Tools
DR is an iron for desoldering jobs of through-hole components and removing excess solder after SMD rework. When the solder container is made of glass you can see when it needs cleaning and the filters need replacing. Some tips are specially designed for cleaning traces of tin from PADs.
  • Capable of providing up to 160 W
  • Very easy cleaning and maintenance
  • Tips are easily placed by thread
Two different types of solder residue chambers are available: metal and glass.