Distilling Head Short Path, Chemglass
Supplier: Chemglass
CG-1239-A-03 CG-1239-A-01
80067-598EA 935.6 USD
80067-598 80067-594
Distilling Head Short Path, Chemglass
Distillation Components Distillation Heads
The condenser is a combination of a cold coil and Liebig.
  • Top outer joint is a 10/30 for use with a 2" immersion thermometer
  • Available with or without Vigreux indentations
Water flows into hose connection “A” through the coil and exits the outer jacket through hose connection “B”.
The head is effective for use distilling medium to high boiling point solvents.