C420 Cartridge Tips for HT420 Thermal Tweezers, JBC
Supplier: JBC TOOLS USA INC. TE
C420271 C420273 C420272
76240-784EA 154.96 USD
76240-784 76241-382 76267-534
C420 Cartridge Tips for HT420 Thermal Tweezers, JBC
Soldering Supplies Soldering Tips
These cartridges are specially appropiate for desoldering chip components and Dual In-line packages.
  • C420 cartridges are appropriate for soldering and desoldering SMD components, reducing handling time
This range works with HT420 Thermal Tweezers which enables aligning catridges and user needs.