Sodium metabisulfite NF, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
di-Sodium disulfite
, Disodiumdisulphite
, Disodium disulfite
, Sodium disulphite
, Disodium disulphite
, Sodium pyrosulphite
, Sodium disulfite
, Disodiumdisulfite
, di-Sodium disulphite
, Sodium pyrosulfite
Management of Change (MOC) category = R
BAKR3550-05
BAKR3550-05
JT3550-5CS
1376.78
USD
JT3550-5
CAJT3550-5
Sodium metabisulfite NF, J.T.Baker®
Sodium metabisulphite
Formula:
Na₂S₂O₅ MW: 190.11 g/mol Density: 1.48 g/cm³ |
MDL Number:
MFCD00167602 CAS Number: 7681-57-4 |
Specification Test Results
GMP Manufactured Product | |
Meets N.F. Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
NF - Identification A | Passes Test |
NF - Identification B | Passes Test |
NF - Limit of Chloride | ≤0.05 % |
NF - Iron (Fe) | ≤0.002 % |
NF - Limit of Thiosulfate | ≤0.05 % |
NF - Assay (SO₂) | 65.0 - 67.4 % |
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...