Qualatrile® 5BQF09 Qualatrile Blue Nitrile 9" Powder-Free Gloves, QRP
Supplier: PIP
BQF09XL BQF09-2X
76242-296CS 418.27 USD
76242-296 76243-420
Qualatrile® 5BQF09 Qualatrile Blue Nitrile 9" Powder-Free Gloves, QRP
Gloves
The QRP 5BQF09 5mil Qualatrile Blue Nitrile 9" Powder-Free Gloves are naturally anti-static with a surface resistance of 10⁹ ohms.
  • Rugged 5 mil thickness
  • 3x puncture resistance of vinyl or natural rubber latex gloves
  • Naturally Anti-static, surface resistance 10⁹ ohms
  • Geogrip 360sm fully textured fingers and palms
RoHS Compliant, 9" Qualatrile Blue 8 mil Powder-Free nitrile ambidextrous gloves are the professional's choice for exceptional personnel, product and process protection.
Beaded cuff for added strength and easier donning. Textured for enhanced wet/dry grip without compromising tactile sensitivity. Manufactured in FDA-registered facilities utilizing an FDA-registered 510(k) formulation. Meets US government 21-CFR specifications; acceptable for use in food and pharmaceutical applications.
Inherently anti-static Qualatrile Blue Nitrile features superior snag and puncture resistance plus excellent flexibility and tactile sensitivity, while protecting the wearer from a broad range of common chemicals.
Certifications: Meets ASTM D6319.