Qualatrile®XC 9C Nitrile Latex-Free Fingercots, QRP
Supplier: PIP
9CM 9CS 9CXL
76238-402CS 2504.81 USD
76238-402 76238-404 76238-406
Qualatrile®XC 9C Nitrile Latex-Free Fingercots, QRP
Finger Cots Nitrile Finger Cots
The nitrile latex-free finger cots have an average surface resistance of 5×10⁹ ohms per square unit.
  • Packaged for ISO 5 (Class 100) applications
  • Average surface resistance of 10⁹ ohms per square unit
  • For use with Class II static sensitive devices (thresholds above 1000 V)
  • Unique, stand-up pouch - easy product identification